siRNA Details
| Gene Symbol: ZNF577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135590.1 | NP_001129062.1 | 7 | 1371 | ||
| NM_032679.2 | NP_116068.2 | 7 | 1524 | |||
| XM_006723432.3 | XP_006723495.1 | 7 | 1548 | |||
| XM_006723433.2 | XP_006723496.1 | 7 | 1551 | |||
| XM_011527408.2 | XP_011525710.1 | 6 | 2507 | |||
| XM_017027387.1 | XP_016882876.1 | 6 | 2330 | |||
| GenBank mRNA | AK289479.1 | - | 7 | 1499 | ||
| AK291812.1 | - | 7 | 1371 | |||
| AL832871.1 | - | 6 | 2289 | |||
| BC004992.1 | - | 7 | 1492 | 2905976 | ||

ZNF577
Genomic Map