siRNA Details
| Gene Symbol: ZNF577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032679.2 | NP_116068.2 | 7 | 694 | ||
| XM_006723432.3 | XP_006723495.1 | 7 | 718 | |||
| XM_006723433.2 | XP_006723496.1 | 7 | 721 | |||
| XM_011527408.2 | XP_011525710.1 | 6 | 1677 | |||
| GenBank mRNA | AK289479.1 | - | 7 | 669 | ||
| BC004992.1 | - | 7 | 662 | 2905976 | ||

ZNF577
Genomic Map