siRNA Details
| Gene Symbol: BUD13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159736.1 | NP_001153208.1 | 9 | 1350 | ||
| NM_032725.3 | NP_116114.1 | 9 | 1752 | |||
| XM_011543035.2 | XP_011541337.1 | 9 | 2326 | |||
| GenBank mRNA | AK057832.1 | - | 9 | 1339 | ||
| AK225164.1 | - | 9 | 1339 | |||
| AK289635.1 | - | 9 | 1741 | |||
| AK291111.1 | - | 9 | 1741 | |||
| BC006350.2 | - | 9 | 1742 | 4111572 | ||
| CA432582.1 | - | 2 | 462 | |||

BUD13
Genomic Map