siRNA Details
| Gene Symbol: KLHL22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032775.3 | NP_116164.2 | 3, 4 | 550 | ||
| XM_017029019.1 | XP_016884508.1 | 3, 4 | 846 | |||
| XM_017029020.1 | XP_016884509.1 | 4, 5 | 2025 | |||
| XM_017029021.1 | XP_016884510.1 | 3, 4 | 479 | |||
| XM_017029025.1 | XP_016884514.1 | 2, 3 | 198 | |||
| GenBank mRNA | AK027266.1 | - | 3, 4 | 489 | ||
| AK222850.1 | - | 3, 4 | 456 | |||
| AK290669.1 | - | 3, 4 | 462 | |||
| BC015923.1 | - | 3, 4 | 502 | 4562420 | ||
| CR456352.1 | - | 2, 3 | 427 | |||

KLHL22
Genomic Map