siRNA Details
| Gene Symbol: ADGRG7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308362.1 | NP_001295291.1 | 8 | 1388 | ||
| NM_032787.2 | NP_116176.2 | 14 | 2212 | |||
| XM_011513245.2 | XP_011511547.1 | 13 | 2087 | |||
| GenBank mRNA | AK027360.1 | - | 14 | 2200 | ||
| AK122804.1 | - | 3 | 1204 | |||
| AK131069.1 | - | 7 | 1071 | |||
| AK172769.1 | - | 10 | 1454 | |||
| AK172774.1 | - | 6 | 965 | |||
| AK295345.1 | - | 8 | 1330 | |||
| AY181246.1 | - | 14 | 1944 | |||
| BC113451.1 | - | 14 | 1978 | 8322503 | ||
| BC130388.1 | - | 14 | 1978 | 40146419 | ||
More Information
Set Membership: |
|

ADGRG7
Genomic Map