siRNA Details
| Gene Symbol: ZNF514 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318005.1 | NP_001304934.1 | 5 | 1147 | ||
| NM_032788.2 | NP_116177.1 | 5 | 767 | |||
| XM_006712806.2 | XP_006712869.1 | 5 | 578 | |||
| GenBank mRNA | AK027363.1 | - | 5 | 700 | ||
| AL832263.1 | - | 3 | 2763 | |||
| BC074900.2 | - | 4 | 361 | 30915480 | ||
| BC074901.2 | - | 4 | 361 | 30915251 | ||
| BC110524.1 | - | 5 | 899 | 40034154 | ||
| BC110525.1 | - | 5 | 519 | 40034155 | ||

ZNF514
Genomic Map