siRNA Details
| Gene Symbol: RNFT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109903.1 | NP_001103373.1 | 7 | 1034 | ||
| NM_032814.3 | NP_116203.2 | 7 | 1034 | |||
| GenBank mRNA | AK027533.1 | - | 8 | 1070 | ||
| AK093164.1 | - | 3 | 835 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNFT2| Gene Symbol: RNFT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109903.1 | NP_001103373.1 | 7 | 1034 | ||
| NM_032814.3 | NP_116203.2 | 7 | 1034 | |||
| GenBank mRNA | AK027533.1 | - | 8 | 1070 | ||
| AK093164.1 | - | 3 | 835 | |||