siRNA Details
| Gene Symbol: CEP89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032816.4 | NP_116205.3 | 4 | 630 | ||
| XM_005259344.3 | XP_005259401.1 | 4 | 625 | |||
| XM_017027398.1 | XP_016882887.1 | 4 | 625 | |||
| GenBank mRNA | AK027546.1 | - | 2 | 223 | ||
| AL832158.1 | - | 4 | 557 | |||
| BC020195.1 | - | 3 | 389 | |||
| BC032307.1 | - | 4 | 564 | |||
| BC136328.1 | - | 4 | 545 | 9020315 | ||

CEP89
Genomic Map