siRNA Details
Gene Symbol: C9orf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193329.1 | NP_001180258.1 | 10 | 2092 | ||
NM_032823.5 | NP_116212.3 | 9 | 1878 | |||
XM_005252284.2 | XP_005252341.1 | 9 | 1943 | |||
XM_006717306.1 | XP_006717369.1 | 11 | 2240 | |||
XM_011519119.1 | XP_011517421.1 | 11 | 2240 | |||
XM_011519120.2 | XP_011517422.1 | 11 | 2240 | |||
XM_011519121.2 | XP_011517423.1 | 11 | 2240 | |||
XM_011519122.2 | XP_011517424.1 | 11 | 2240 | |||
XM_011519123.2 | XP_011517425.1 | 11 | 2240 | |||
XM_011519127.1 | XP_011517429.1 | 10 | 2196 | |||
XM_011519132.1 | XP_011517434.1 | 8 | 966 | |||
XM_017015229.1 | XP_016870718.1 | 11 | 2283 | |||
XM_017015230.1 | XP_016870719.1 | 11 | 2240 | |||
XM_017015231.1 | XP_016870720.1 | 9 | 1943 | |||
XM_017015232.1 | XP_016870721.1 | 11 | 2240 | |||
XM_017015233.1 | XP_016870722.1 | 11 | 2270 | |||
XM_017015234.1 | XP_016870723.1 | 10 | 2073 | |||
XM_017015235.1 | XP_016870724.1 | 10 | 2073 | |||
XM_017015236.1 | XP_016870725.1 | 8 | 1776 | |||
XM_017015237.1 | XP_016870726.1 | 8 | 1776 | |||
GenBank mRNA | AJ560639.1 | - | 10 | 1957 | ||
AK027581.1 | - | 8 | 1129 | |||
BC020194.1 | - | 10 | 1303 | 5020674 | ||
BC029616.1 | - | 9 | 1835 | 3877098 | ||
BX648970.1 | - | 9 | 1178 |
