siRNA Details
| Gene Symbol: ATOH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032827.6 | NP_116216.2 | 2 | 1235 | ||
| XM_006712122.3 | XP_006712185.1 | 2 | 1235 | |||
| XM_011533139.1 | XP_011531441.1 | 2 | 1235 | |||
| GenBank mRNA | AF529205.1 | - | 2 | 831 | ||
| AK027614.1 | - | 2 | 1028 | |||
| AK074681.1 | - | 2 | 1127 | |||
| AK096049.1 | - | 2 | 582 | |||
| AL831857.1 | - | 2 | 374 | |||
| BC021207.2 | - | 2 | 1126 | 4109799 | ||
| BC094832.1 | - | 2 | 905 | 6292270 | ||

ATOH8
Genomic Map