siRNA Details
| Gene Symbol: SPRYD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032840.2 | NP_116229.1 | 9 | 1065 | ||
| GenBank mRNA | AK027706.1 | - | 9 | 999 | ||
| AK074566.1 | - | 4 | 349 | |||
| AK074694.1 | - | 9 | 1031 | |||
| AK294092.1 | - | 8 | 913 | |||
| BC136312.1 | - | 9 | 998 | 9020299 | ||

SPRYD3
Genomic Map