siRNA Details
| Gene Symbol: LINC01101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027181.1 | 1 | 561 | |||
| GenBank mRNA | AK027722.1 | - | 1 | 333 | ||
| BC112205.1 | - | 1 | 441 | |||
| BC113708.1 | - | 1 | 429 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01101| Gene Symbol: LINC01101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027181.1 | 1 | 561 | |||
| GenBank mRNA | AK027722.1 | - | 1 | 333 | ||
| BC112205.1 | - | 1 | 441 | |||
| BC113708.1 | - | 1 | 429 | |||