siRNA Details
| Gene Symbol: LTV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032860.3 | NP_116249.2 | 6 | 739 | ||
| GenBank mRNA | AK027815.1 | - | 4 | 488 | ||
| AK091447.1 | - | 1 | 3788 | |||
| AK300360.1 | - | 5 | 1032 | |||
| BC009855.2 | - | 6 | 739 | 3926227 | ||

LTV1
Genomic Map