siRNA Details
| Gene Symbol: SERAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032861.3 | NP_116250.3 | 14 | 1616 | ||
| NR_073096.1 | 12 | 1407 | ||||
| XM_006715586.2 | XP_006715649.1 | 13 | 1545 | |||
| XM_011536196.2 | XP_011534498.1 | 13 | 1639 | |||
| XM_011536198.2 | XP_011534500.1 | 12 | 1541 | |||
| GenBank mRNA | AK027823.1 | - | 14 | 1606 | ||
| AK125878.1 | - | 15 | 2808 | |||
| BC001705.1 | - | 14 | 1573 | 3350731 | ||
| BX648104.1 | - | 12 | 2992 | |||

SERAC1
Genomic Map