siRNA Details
Gene Symbol: MICALCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032867.2 | NP_116256.2 | 3 | 1413 | ||
GenBank mRNA | AK027872.1 | - | 1 | 731 | ||
AK123671.1 | - | 3 | 1413 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MICALCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032867.2 | NP_116256.2 | 3 | 1413 | ||
GenBank mRNA | AK027872.1 | - | 1 | 731 | ||
AK123671.1 | - | 3 | 1413 |