siRNA Details
| Gene Symbol: MICALCL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032867.2 | NP_116256.2 | 3 | 1585 | ||
| GenBank mRNA | AK027872.1 | - | 1 | 903 | ||
| AK123671.1 | - | 3 | 1585 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MICALCL| Gene Symbol: MICALCL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032867.2 | NP_116256.2 | 3 | 1585 | ||
| GenBank mRNA | AK027872.1 | - | 1 | 903 | ||
| AK123671.1 | - | 3 | 1585 | |||