siRNA Details
| Gene Symbol: UBASH3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032873.4 | NP_116262.2 | 4 | 816 | ||
| XM_005271712.3 | XP_005271769.1 | 4 | 3062 | |||
| XM_011543041.2 | XP_011541343.1 | 4 | 454 | |||
| XM_011543042.1 | XP_011541344.1 | 4 | 820 | |||
| GenBank mRNA | AB075839.1 | - | 4 | 668 | ||
| AF425252.1 | - | 2 | 113 | |||
| AK075203.1 | - | 4 | 784 | |||
| AK222843.1 | - | 4 | 766 | |||
| AK222846.1 | - | 4 | 766 | |||
| BC007541.1 | - | 4 | 770 | 2958242 | ||

UBASH3B
Genomic Map