siRNA Details
| Gene Symbol: FBXL20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184906.1 | NP_001171835.1 | 4 | 458 | ||
| NM_032875.2 | NP_116264.2 | 4 | 458 | |||
| XM_005257746.3 | XP_005257803.2 | 4 | 649 | |||
| XM_005257747.3 | XP_005257804.2 | 4 | 526 | |||
| GenBank mRNA | AK123048.1 | - | 4 | 459 | ||
| AK291844.1 | - | 4 | 469 | |||
| AK312612.1 | - | 4 | 439 | |||
| BC007557.1 | - | 4 | 425 | 2987858 | ||
| DA747594.1 | - | 4 | 458 | |||
| DQ223959.1 | - | 4 | 425 | |||

FBXL20
Genomic Map