siRNA Details
| Gene Symbol: FBXL20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032875.2 | NP_116264.2 | 7 | 702 | ||
| XM_005257746.3 | XP_005257803.2 | 7 | 893 | |||
| GenBank mRNA | AK123048.1 | - | 7 | 703 | ||
| AK291844.1 | - | 7 | 713 | |||
| AK312612.1 | - | 7 | 683 | |||
| BC007557.1 | - | 7 | 669 | 2987858 | ||

FBXL20
Genomic Map