siRNA Details
| Gene Symbol: LSM10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032881.2 | NP_116270.1 | 2 | 240 | ||
| GenBank mRNA | AF166092.1 | - | 2 | 166 | ||
| AF394685.1 | - | 2 | 138 | |||
| BC007623.1 | - | 2 | 173 | 3357043 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LSM10| Gene Symbol: LSM10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032881.2 | NP_116270.1 | 2 | 240 | ||
| GenBank mRNA | AF166092.1 | - | 2 | 166 | ||
| AF394685.1 | - | 2 | 138 | |||
| BC007623.1 | - | 2 | 173 | 3357043 | ||