siRNA Details
| Gene Symbol: TMEM128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297551.1 | NP_001284480.1 | 2 | 173 | ||
| NM_001297552.1 | NP_001284481.1 | 2 | 173 | |||
| NM_032927.3 | NP_116316.1 | 2 | 678 | |||
| XM_005248034.2 | XP_005248091.1 | 2 | 196 | |||
| GenBank mRNA | AK295248.1 | - | 2 | 155 | ||
| AK296007.1 | - | 2 | 154 | |||
| BC007729.2 | - | 2 | 643 | 4303698 | ||
| BC091482.1 | - | 2 | 173 | 30344390 | ||
| BM263799.1 | - | 2 | 155 | |||
| DA019506.1 | - | 2 | 163 | |||

TMEM128
Genomic Map