siRNA Details
| Gene Symbol: ATCAY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033064.4 | NP_149053.1 | 2 | 488 | ||
| GenBank mRNA | AB058775.1 | - | 2 | 434 | ||
| AK027889.1 | - | 2 | 423 | |||
| AK125457.1 | - | 2 | 131 | |||
| AK299522.1 | - | 2 | 423 | |||
| AY220297.1 | - | 1 | 58 | |||
| BC026217.1 | - | 2 | 430 | 4153341 | ||
| DA151785.1 | - | 2 | 467 | |||
| DA778097.1 | - | 2 | 488 | |||
| KU178814.1 | - | 1 | 58 | |||
| KU178815.1 | - | 1 | 58 | |||

ATCAY
Genomic Map