siRNA Details
| Gene Symbol: ITPRIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001272012.1 | NP_001258941.1 | 2 | 1145 | ||
| NM_001272013.1 | NP_001258942.1 | 2 | 1265 | |||
| NM_033397.3 | NP_203755.1 | 3 | 1457 | |||
| XM_005270257.2 | XP_005270314.1 | 1 | 1021 | |||
| GenBank mRNA | AB051541.1 | - | 2 | 1176 | ||
| AK056526.1 | - | 2 | 1077 | |||
| AK094802.1 | - | 1 | 497 | |||
| AK129877.1 | - | 3 | 1168 | |||
| BC040536.1 | - | 2 | 1117 | 5265379 | ||
| BC070108.1 | - | 3 | 1457 | 4837204 | ||

ITPRIP
Genomic Map