siRNA Details
| Gene Symbol: FHDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033393.2 | NP_203751.2 | 8 | 1162 | ||
| XM_005263319.3 | XP_005263376.1 | 9 | 1303 | |||
| XM_006714393.3 | XP_006714456.1 | 9 | 1312 | |||
| XM_011532388.1 | XP_011530690.1 | 11 | 1794 | |||
| XM_011532389.1 | XP_011530691.1 | 9 | 1908 | |||
| XM_017008787.1 | XP_016864276.1 | 7 | 1147 | |||
| GenBank mRNA | AB051514.1 | - | 8 | 1049 | ||

FHDC1
Genomic Map