siRNA Details
| Gene Symbol: EPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB051511.1 | - | 3 | 288 | ||
| AB621826.1 | - | 3 | 281 | |||
| AK095620.1 | - | 3 | 298 | |||
| BC021229.2 | - | 3 | 243 | 4111329 | ||
| DA619246.1 | - | 3 | 326 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EPT1| Gene Symbol: EPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB051511.1 | - | 3 | 288 | ||
| AB621826.1 | - | 3 | 281 | |||
| AK095620.1 | - | 3 | 298 | |||
| BC021229.2 | - | 3 | 243 | 4111329 | ||
| DA619246.1 | - | 3 | 326 | |||