siRNA Details
| Gene Symbol: PSKH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033126.1 | NP_149117.1 | 1, 2 | 184 | ||
| GenBank mRNA | AY037806.1 | - | 1, 2 | 184 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PSKH2| Gene Symbol: PSKH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033126.1 | NP_149117.1 | 1, 2 | 184 | ||
| GenBank mRNA | AY037806.1 | - | 1, 2 | 184 | ||