siRNA Details
| Gene Symbol: GTPBP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042717.2 | NP_001036182.1 | 7 | 704 | ||
| NM_033107.3 | NP_149098.2 | 9 | 941 | |||
| GenBank mRNA | AF351613.1 | - | 7 | 705 | ||
| AK095561.1 | - | 9 | 931 | |||
| AK294325.1 | - | 7 | 654 | |||
| BC004923.2 | - | 8 | 838 | |||
| BC021573.1 | - | 8 | 838 | |||
| BC107714.1 | - | 9 | 925 | 6714887 | ||
| BC144162.1 | - | 7 | 651 | |||
| CR933597.1 | - | 8 | 755 | |||

GTPBP10
Genomic Map