siRNA Details
| Gene Symbol: GAL3ST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033036.2 | NP_149025.1 | 3 | 526 | ||
| XM_017018519.1 | XP_016874008.1 | 2 | 440 | |||
| GenBank mRNA | AB053232.1 | - | 3 | 526 | ||
| AY026481.1 | - | 2 | 404 | |||
| BC113552.1 | - | 2 | 312 | 8322604 | ||
| BC113554.1 | - | 2 | 312 | 8322606 | ||

GAL3ST3
Genomic Map