siRNA Details
| Gene Symbol: KLHL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130446.2 | NP_569713.2 | 1 | 171 | ||
| XM_011513274.2 | XP_011511576.1 | 1 | 172 | |||
| GenBank mRNA | AF441792.1 | - | 1 | 83 | ||
| AK097125.1 | - | 1 | 125 | |||
| AK314473.1 | - | 1 | 125 | |||
| BC032348.1 | - | 1 | 149 | 5213743 | ||
| BP284594.1 | - | 1 | 171 | |||
| BX648370.1 | - | 1 | 126 | |||

KLHL6
Genomic Map