siRNA Details
| Gene Symbol: SEC16B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033127.2 | NP_149118.2 | 21 | 2740 | ||
| GenBank mRNA | AB063357.1 | - | 21 | 2740 | ||
| AK074451.1 | - | 2 | 158 | |||
| AK093939.1 | - | 9 | 981 | |||
| AK098627.1 | - | 10 | 1360 | |||
| AK314585.1 | - | 20 | 2744 | |||
| EF125213.1 | - | 20 | 2650 | |||

SEC16B
Genomic Map