siRNA Details
| Gene Symbol: WDR34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052844.3 | NP_443076.2 | 8 | 1430 | ||
| XM_011519179.2 | XP_011517481.1 | 8 | 1280 | |||
| GenBank mRNA | BC001614.1 | - | 8 | 1265 | ||
| BC011874.2 | - | 8 | 1367 | 3161070 | ||

WDR34
Genomic Map