siRNA Details
| Gene Symbol: C16orf45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033201.2 | NP_149978.1 | 1, 2 | 296 | ||
| XM_011522731.2 | XP_011521033.1 | 1, 2 | 275 | |||
| XM_017023824.1 | XP_016879313.1 | 1, 2 | 273 | |||
| GenBank mRNA | AK057180.1 | - | 1, 2 | 296 | ||
| BC008967.2 | - | 1, 2 | 226 | 3010666 | ||

C16orf45
Genomic Map