siRNA Details
| Gene Symbol: CCDC120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163321.2 | NP_001156793.2 | 10 | 1911 | ||
| NM_001163322.2 | NP_001156794.1 | 10 | 1852 | |||
| NM_001163323.2 | NP_001156795.1 | 10 | 1888 | |||
| NM_001271835.1 | NP_001258764.1 | 10 | 1888 | |||
| NM_001271836.1 | NP_001258765.1 | 10 | 2240 | |||
| NM_033626.3 | NP_296375.1 | 10 | 1829 | |||
| GenBank mRNA | AK056290.1 | - | 1 | 425 | ||
| AK125424.1 | - | 1 | 1423 | |||
| AK291370.1 | - | 10 | 2100 | |||
| AK293898.1 | - | 10 | 1797 | |||
| AK294024.1 | - | 10 | 1875 | |||
| AK300366.1 | - | 10 | 1839 | |||
| AK315837.1 | - | 10 | 1855 | |||
| BC008769.2 | - | 10 | 1810 | 3139519 | ||

CCDC120
Genomic Map