siRNA Details
| Gene Symbol: OTULIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138348.4 | NP_612357.4 | 6 | 829 | ||
| XM_011514151.2 | XP_011512453.1 | 6 | 4709 | |||
| XM_011514152.2 | XP_011512454.1 | 6 | 4709 | |||
| XM_017010015.1 | XP_016865504.1 | 5 | 4632 | |||
| GenBank mRNA | AK092203.1 | - | 2 | 149 | ||
| BC007706.2 | - | 4 | 402 | 3952677 | ||
| BC015392.1 | - | 6 | 755 | 4430188 | ||

OTULIN
Genomic Map