siRNA Details
| Gene Symbol: IFT20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267774.1 | NP_001254703.1 | 4 | 549 | ||
| NM_001267775.1 | NP_001254704.1 | 4 | 348 | |||
| NM_001267776.1 | NP_001254705.1 | 3 | 312 | |||
| NM_001267777.1 | NP_001254706.1 | 4 | 348 | |||
| NM_001267778.1 | NP_001254707.1 | 3 | 312 | |||
| NM_174887.3 | NP_777547.1 | 3 | 312 | |||
| GenBank mRNA | AA490722.1 | - | 3 | 228 | ||
| AK297769.1 | - | 4 | 515 | |||
| AY224601.1 | - | 4 | 498 | |||
| BC002640.2 | - | 3 | 292 | 3609552 | ||
| BC038094.1 | - | 3 | 278 | 3907361 | ||
| BE619475.1 | - | 4 | 284 | |||
| BQ639919.1 | - | 4 | 298 | |||

IFT20
Genomic Map