siRNA Details
| Gene Symbol: SCRN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145023.1 | NP_001138495.1 | 4 | 526 | ||
| NM_138355.3 | NP_612364.2 | 4 | 526 | |||
| XM_005257776.4 | XP_005257833.1 | 4 | 506 | |||
| XM_017025286.1 | XP_016880775.1 | 4 | 454 | |||
| GenBank mRNA | AK290646.1 | - | 4 | 429 | ||
| AK303835.1 | - | 4 | 482 | |||
| BC002980.1 | - | 4 | 469 | |||
| BC010408.2 | - | 4 | 440 | 3945715 | ||
| BC017317.2 | - | 4 | 439 | 3832079 | ||

SCRN2
Genomic Map