siRNA Details
Gene Symbol: YIF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039671.2 | NP_001034760.1 | 6 | 707 | ||
NM_001039672.2 | NP_001034761.1 | 6 | 725 | |||
NM_001039673.2 | NP_001034762.1 | 6 | 716 | |||
NM_001145461.1 | NP_001138933.1 | 6 | 701 | |||
NM_001145462.1 | NP_001138934.1 | 6 | 677 | |||
NM_001145463.1 | NP_001138935.1 | 6 | 716 | |||
XM_005259385.3 | XP_005259442.1 | 6 | 938 | |||
XM_017027449.1 | XP_016882938.1 | 6 | 726 | |||
XM_017027450.1 | XP_016882939.1 | 6 | 724 | |||
XM_017027451.1 | XP_016882940.1 | 6 | 722 | |||
GenBank mRNA | AF258578.3 | - | 5 | 899 | ||
AK304479.1 | - | 3 | 1063 | |||
AL833382.1 | - | 6 | 673 | |||
AY358965.1 | - | 5 | 844 | |||
BC007644.1 | - | 6 | 664 | |||
BC014974.1 | - | 6 | 670 | 4865479 | ||
BC091477.1 | - | 6 | 699 | 30344388 | ||
BX379501.2 | - | 6 | 643 | |||
CA487891.1 | - | 6 | 659 | |||
CA488303.1 | - | 6 | 654 |
