siRNA Details
| Gene Symbol: ZNF439 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152262.2 | NP_689475.1 | 1 | 145 | ||
| GenBank mRNA | AL833935.1 | - | 1 | 146 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF439| Gene Symbol: ZNF439 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152262.2 | NP_689475.1 | 1 | 145 | ||
| GenBank mRNA | AL833935.1 | - | 1 | 146 | ||