siRNA Details
| Gene Symbol: ZNF439 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152262.2 | NP_689475.1 | 3 | 1486 | ||
| GenBank mRNA | AK027542.1 | - | 1 | 1113 | ||
| AL833935.1 | - | 3 | 1487 | |||
| BC032857.1 | - | 3 | 1358 | 5268314 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF439| Gene Symbol: ZNF439 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152262.2 | NP_689475.1 | 3 | 1486 | ||
| GenBank mRNA | AK027542.1 | - | 1 | 1113 | ||
| AL833935.1 | - | 3 | 1487 | |||
| BC032857.1 | - | 3 | 1358 | 5268314 | ||