siRNA Details
| Gene Symbol: TBL1Y | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033284.1 | NP_150600.1 | 15 | 1748 | ||
| NM_134258.1 | NP_599020.1 | 14 | 1717 | |||
| NM_134259.1 | NP_599021.1 | 14 | 1653 | |||
| XM_005262572.3 | XP_005262629.1 | 9 | 1412 | |||
| XM_017030086.1 | XP_016885575.1 | 15 | 1737 | |||
| XM_017030087.1 | XP_016885576.1 | 15 | 1737 | |||
| GenBank mRNA | AF332220.1 | - | 15 | 1748 | ||
| AF332221.1 | - | 14 | 1717 | |||
| AF332222.1 | - | 14 | 1653 | |||
| AK298210.1 | - | 15 | 1763 | |||
| BC130471.1 | - | 11 | 1231 | 40146502 | ||
| BC130473.1 | - | 11 | 1231 | 40146504 | ||

TBL1Y
Genomic Map