siRNA Details
| Gene Symbol: CARMIL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138360.3 | NP_612369.3 | 37, 38 | 3947 | ||
| XM_011537300.2 | XP_011535602.1 | 37, 38 | 3738 | |||
| XM_017021751.1 | XP_016877240.1 | 34, 35 | 3639 | |||
| GenBank mRNA | AL834197.1 | - | 8, 9 | 2015 | ||
| BC008134.2 | - | 4, 5 | 394 | |||

CARMIL3
Genomic Map