siRNA Details
| Gene Symbol: CCDC126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138771.3 | NP_620126.2 | 3 | 589 | ||
| XM_017012775.1 | XP_016868264.1 | 4 | 829 | |||
| XM_017012776.1 | XP_016868265.1 | 3 | 572 | |||
| GenBank mRNA | AK026684.1 | - | 1 | 86 | ||
| AK289911.1 | - | 3 | 565 | |||
| AY358713.1 | - | 3 | 556 | |||
| BC012427.1 | - | 1 | 170 | 3858521 | ||
| BC104638.1 | - | 3 | 481 | 6620343 | ||

CCDC126
Genomic Map