siRNA Details
| Gene Symbol: PRMT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304458.1 | NP_001291387.1 | 7 | 1311 | ||
| NM_138364.3 | NP_612373.2 | 8 | 1460 | |||
| XM_017008823.1 | XP_016864312.1 | 8 | 1201 | |||
| XM_017008824.1 | XP_016864313.1 | 7 | 1133 | |||
| XM_017008825.1 | XP_016864314.1 | 7 | 1187 | |||
| XM_017008826.1 | XP_016864315.1 | 6 | 1082 | |||
| GenBank mRNA | AK096703.1 | - | 7 | 1206 | ||
| AK128483.1 | - | 2 | 543 | |||
| AK292904.1 | - | 8 | 1393 | |||
| BC021250.2 | - | 8 | 1215 | |||
| BC064403.1 | - | 8 | 1359 | 5547611 | ||

PRMT9
Genomic Map