siRNA Details
| Gene Symbol: PRMT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304458.1 | NP_001291387.1 | 8 | 1900 | ||
| NM_138364.3 | NP_612373.2 | 9 | 2049 | |||
| XM_017008823.1 | XP_016864312.1 | 9 | 1790 | |||
| XM_017008824.1 | XP_016864313.1 | 8 | 1722 | |||
| XM_017008825.1 | XP_016864314.1 | 8 | 1776 | |||
| XM_017008826.1 | XP_016864315.1 | 7 | 1671 | |||
| GenBank mRNA | AK096703.1 | - | 8 | 1795 | ||
| AK128483.1 | - | 3 | 1132 | |||
| AK292904.1 | - | 9 | 1982 | |||
| BC004337.1 | - | 1 | 328 | |||
| BC021250.2 | - | 9 | 1804 | |||
| BC064403.1 | - | 9 | 1948 | 5547611 | ||

PRMT9
Genomic Map