siRNA Details
| Gene Symbol: PRMT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304458.1 | NP_001291387.1 | 10 | 2324 | ||
| NM_138364.3 | NP_612373.2 | 11 | 2473 | |||
| XM_017008823.1 | XP_016864312.1 | 11 | 2214 | |||
| XM_017008824.1 | XP_016864313.1 | 10 | 2146 | |||
| XM_017008825.1 | XP_016864314.1 | 10 | 2200 | |||
| XM_017008826.1 | XP_016864315.1 | 9 | 2095 | |||
| GenBank mRNA | AK096703.1 | - | 10 | 2219 | ||
| AK128483.1 | - | 5 | 1556 | |||
| AK292904.1 | - | 11 | 2406 | |||
| AL137452.1 | - | 2 | 199 | |||
| BC021250.2 | - | 11 | 2228 | |||
| BC064403.1 | - | 11 | 2372 | 5547611 | ||

PRMT9
Genomic Map