siRNA Details
| Gene Symbol: AP5B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138368.4 | NP_612377.4 | 2 | 1665 | ||
| GenBank mRNA | AF193040.1 | - | 1 | 1016 | ||
| AL834269.1 | - | 1 | 562 | |||
| BC109109.1 | - | 1 | 1198 | 40035346 | ||
| BC109110.1 | - | 1 | 1198 | 40035347 | ||
| BC126905.1 | - | 1 | 1198 | 40035350 | ||
| BC128056.1 | - | 1 | 1198 | 40035353 | ||
| JQ313135.1 | - | 2 | 1665 | |||

AP5B1
Genomic Map