siRNA Details
| Gene Symbol: TIGD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033208.3 | NP_149985.2 | 2 | 2368 | ||
| GenBank mRNA | AF251050.2 | - | 1 | 1531 | ||
| AK308054.1 | - | 1 | 1508 | |||
| BC069623.1 | - | 1 | 987 | 7262458 | ||
| BC069632.1 | - | 1 | 986 | 7262470 | ||

TIGD7
Genomic Map