siRNA Details
| Gene Symbol: CCDC150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006712439.3 | XP_006712502.1 | 1, 2 | 589 | ||
| GenBank mRNA | AK125768.1 | - | 1, 2 | 589 | ||
| AL834537.1 | - | 1, 2 | 484 | |||
| BC062658.1 | - | 1, 2 | 545 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC150| Gene Symbol: CCDC150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006712439.3 | XP_006712502.1 | 1, 2 | 589 | ||
| GenBank mRNA | AK125768.1 | - | 1, 2 | 589 | ||
| AL834537.1 | - | 1, 2 | 484 | |||
| BC062658.1 | - | 1, 2 | 545 | |||