siRNA Details
| Gene Symbol: LONRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152271.3 | NP_689484.3 | 7 | 1604 | ||
| XM_005273685.3 | XP_005273742.1 | 7 | 1573 | |||
| XM_011544694.2 | XP_011542996.1 | 7 | 1356 | |||
| GenBank mRNA | AK074329.1 | - | 2 | 156 | ||
| AK297266.1 | - | 6 | 741 | |||
| AK300538.1 | - | 3 | 350 | |||
| BC106042.1 | - | 7 | 870 | |||
| BC117381.1 | - | 4 | 535 | 40125932 | ||
| BC117385.1 | - | 4 | 535 | 40125936 | ||
| BQ718281.1 | - | 201 | ||||

LONRF1
Genomic Map