siRNA Details
| Gene Symbol: ELMSAN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001043318.2 | NP_001036783.1 | 10 | 3109 | ||
| NM_194278.3 | NP_919254.2 | 10 | 3488 | |||
| XM_005268204.1 | XP_005268261.1 | 10 | 3488 | |||
| XM_005268205.1 | XP_005268262.1 | 10 | 3044 | |||
| XM_005268206.1 | XP_005268263.1 | 11 | 3261 | |||
| GenBank mRNA | AK127785.1 | - | 11 | 3487 | ||
| BC003121.1 | - | 10 | 2281 | |||
| BC006451.1 | - | 10 | 1943 | |||
| BC006511.2 | - | 10 | 2739 | |||
| BC009202.2 | - | 11 | 3109 | |||
| BC015668.2 | - | 10 | 3122 | |||
| BC025330.1 | - | 10 | 2739 | |||

ELMSAN1
Genomic Map